Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 163970 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC6A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLC6A2 - solute carrier family 6 member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001043.3 | 3 | Intron | NP_001034.1 | |||
NM_001172501.1 | 3 | UTR 5 | NP_001165972.1 | |||
NM_001172502.1 | 3 | Intron | NP_001165973.1 | |||
NM_001172504.1 | 3 | Intron | NP_001165975.1 | |||
XM_006721263.2 | 3 | Intron | XP_006721326.1 | |||
XM_011523295.1 | 3 | UTR 5 | XP_011521597.1 | |||
XM_011523296.1 | 3 | UTR 5 | XP_011521598.1 | |||
XM_011523297.2 | 3 | UTR 5 | XP_011521599.1 | |||
XM_011523298.1 | 3 | UTR 5 | XP_011521600.1 | |||
XM_011523299.2 | 3 | Intron | XP_011521601.1 | |||
XM_011523300.2 | 3 | Intron | XP_011521602.1 |
Set Membership: |
HapMap |