Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 103830 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKR1A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AKR1A1 - aldo-keto reductase family 1 member A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202413.1 | Intron | NP_001189342.1 | ||||
NM_001202414.1 | Intron | NP_001189343.1 | ||||
NM_006066.3 | Intron | NP_006057.1 | ||||
NM_153326.2 | Intron | NP_697021.1 | ||||
XM_011540491.2 | Intron | XP_011538793.1 | ||||
XM_011540492.2 | Intron | XP_011538794.1 |