Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FER1L5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FER1L5 - fer-1 like family member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001293083.1 | Intron | NP_001280012.1 | ||||
XM_011512110.2 | Intron | XP_011510412.1 | ||||
XM_011512111.2 | Intron | XP_011510413.1 | ||||
XM_011512113.2 | Intron | XP_011510415.1 | ||||
XM_011512114.2 | Intron | XP_011510416.1 | ||||
XM_011512115.2 | Intron | XP_011510417.1 | ||||
XM_011512116.2 | Intron | XP_011510418.1 | ||||
XM_011512117.2 | Intron | XP_011510419.1 | ||||
XM_011512118.2 | Intron | XP_011510420.1 | ||||
XM_011512119.2 | Intron | XP_011510421.1 | ||||
XM_011512120.2 | Intron | XP_011510422.1 | ||||
XM_011512122.2 | Intron | XP_011510424.1 | ||||
XM_011512124.2 | Intron | XP_011510426.1 | ||||
XM_011512125.2 | Intron | XP_011510427.1 | ||||
XM_011512126.2 | Intron | XP_011510428.1 | ||||
XM_011512127.2 | Intron | XP_011510429.1 | ||||
XM_011512128.2 | Intron | XP_011510430.1 | ||||
XM_017005222.1 | Intron | XP_016860711.1 | ||||
XM_017005223.1 | Intron | XP_016860712.1 | ||||
XM_017005224.1 | Intron | XP_016860713.1 |
KANSL3 - KAT8 regulatory NSL complex subunit 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |