Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603728 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUMB PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NUMB - NUMB, endocytic adaptor protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005743.1 | 2162 | Silent Mutation | AAC,AAT | N,N 625 | NP_001005743.1 | |
NM_001005744.1 | 2162 | Silent Mutation | AAC,AAT | N,N 577 | NP_001005744.1 | |
NM_001005745.1 | 2162 | Silent Mutation | AAC,AAT | N,N 566 | NP_001005745.1 | |
NM_001320114.1 | 2162 | Silent Mutation | AAC,AAT | N,N 577 | NP_001307043.1 | |
NM_003744.5 | 2162 | Silent Mutation | AAC,AAT | N,N 614 | NP_003735.3 |
PAPLN - papilin, proteoglycan like sulfated glycoprotein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |