Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCAGCTCATTCCGGCTTCACGCCA[C/T]GTCGGGCAGTTCACGGTTTGGGTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101928123 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC101928123 - uncharacterized LOC101928123 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PAPLN - papilin, proteoglycan like sulfated glycoprotein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173462.3 | Intron | NP_775733.3 | ||||
XM_011537290.2 | Intron | XP_011535592.1 | ||||
XM_011537291.2 | Intron | XP_011535593.1 | ||||
XM_011537292.2 | Intron | XP_011535594.1 | ||||
XM_011537293.2 | Intron | XP_011535595.1 | ||||
XM_011537294.2 | Intron | XP_011535596.1 | ||||
XM_011537295.2 | Intron | XP_011535597.1 | ||||
XM_011537296.2 | Intron | XP_011535598.1 | ||||
XM_017021749.1 | Intron | XP_016877238.1 | ||||
XM_017021750.1 | Intron | XP_016877239.1 |
Set Membership: |
HapMap |