Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTTCGCGTACAGGAGAAGAAACA[C/G]GGATGATCTCTGTCTTTATTTTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605165 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PATZ1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
PATZ1 - POZ/BTB and AT hook containing zinc finger 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014323.2 | Intron | NP_055138.2 | ||||
NM_032050.1 | Intron | NP_114439.1 | ||||
NM_032051.1 | Intron | NP_114440.1 | ||||
NM_032052.1 | Intron | NP_114441.1 |
PIK3IP1-AS1 - PIK3IP1 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |