Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601142 MIM: 607215 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNAB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCNAB2 - potassium voltage-gated channel subfamily A regulatory beta subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199860.1 | Intron | NP_001186789.1 | ||||
NM_001199861.1 | Intron | NP_001186790.1 | ||||
NM_001199862.1 | Intron | NP_001186791.1 | ||||
NM_001199863.1 | Intron | NP_001186792.1 | ||||
NM_003636.3 | Intron | NP_003627.1 | ||||
NM_172130.2 | Intron | NP_742128.1 | ||||
XM_005263514.2 | Intron | XP_005263571.1 | ||||
XM_011542321.2 | Intron | XP_011540623.1 | ||||
XM_011542322.2 | Intron | XP_011540624.1 | ||||
XM_017002618.1 | Intron | XP_016858107.1 | ||||
XM_017002619.1 | Intron | XP_016858108.1 | ||||
XM_017002620.1 | Intron | XP_016858109.1 | ||||
XM_017002621.1 | Intron | XP_016858110.1 |
NPHP4 - nephronophthisis 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |