Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGACTCGGCATTGGGAAGCTGGAG[T/C]CACACCTGGTCATTTTCTGTGAGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 120110 | ||||||||||||||||||||
Literature Links: |
COL10A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COL10A1 - collagen type X alpha 1 chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000493.3 | 5496 | Nonsense Mutation | TGA,TGG | *,W 651 | NP_000484.2 | |
XM_006715333.3 | 5496 | Nonsense Mutation | TGA,TGG | *,W 651 | XP_006715396.1 | |
XM_011535432.2 | 5496 | Nonsense Mutation | TGA,TGG | *,W 651 | XP_011533734.1 | |
XM_011535433.2 | 5496 | Nonsense Mutation | TGA,TGG | *,W 651 | XP_011533735.1 | |
XM_017010248.1 | 5496 | Nonsense Mutation | TGA,TGG | *,W 651 | XP_016865737.1 | |
XM_017010249.1 | 5496 | Nonsense Mutation | TGA,TGG | *,W 651 | XP_016865738.1 |
NT5DC1 - 5'-nucleotidase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152729.2 | 5496 | Intron | NP_689942.2 | |||
XM_006715378.3 | 5496 | Intron | XP_006715441.1 |