Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF568 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF568 - zinc finger protein 568 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204835.1 | Intron | NP_001191764.1 | ||||
NM_001204836.1 | Intron | NP_001191765.1 | ||||
NM_001204837.1 | Intron | NP_001191766.1 | ||||
NM_001204838.1 | Intron | NP_001191767.1 | ||||
NM_001204839.1 | Intron | NP_001191768.1 | ||||
NM_198539.3 | Intron | NP_940941.2 | ||||
XM_017026772.1 | Intron | XP_016882261.1 | ||||
XM_017026773.1 | Intron | XP_016882262.1 | ||||
XM_017026774.1 | Intron | XP_016882263.1 | ||||
XM_017026775.1 | Intron | XP_016882264.1 | ||||
XM_017026776.1 | Intron | XP_016882265.1 | ||||
XM_017026777.1 | Intron | XP_016882266.1 | ||||
XM_017026778.1 | Intron | XP_016882267.1 | ||||
XM_017026779.1 | Intron | XP_016882268.1 | ||||
XM_017026780.1 | Intron | XP_016882269.1 |
ZNF829 - zinc finger protein 829 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |