Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 165230 | ||||||||||||||||||||
Literature Links: |
GLI2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GLI2 - GLI family zinc finger 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005270.4 | Intron | NP_005261.2 | ||||
XM_006712422.2 | Intron | XP_006712485.1 | ||||
XM_011510969.2 | Intron | XP_011509271.2 | ||||
XM_011510970.2 | Intron | XP_011509272.1 | ||||
XM_011510971.2 | Intron | XP_011509273.1 | ||||
XM_011510972.2 | Intron | XP_011509274.2 | ||||
XM_011510973.2 | Intron | XP_011509275.1 | ||||
XM_011510974.2 | Intron | XP_011509276.1 | ||||
XM_017003818.1 | Intron | XP_016859307.1 |