Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC130 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC130 - coiled-coil domain containing 130 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320561.1 | Intron | NP_001307490.1 | ||||
NM_001320564.1 | Intron | NP_001307493.1 | ||||
NM_001320565.1 | Intron | NP_001307494.1 | ||||
NM_001320566.1 | Intron | NP_001307495.1 | ||||
NM_001320567.1 | Intron | NP_001307496.1 | ||||
NM_001320568.1 | Intron | NP_001307497.1 | ||||
NM_001320569.1 | Intron | NP_001307498.1 | ||||
NM_030818.3 | Intron | NP_110445.1 | ||||
XM_005260086.4 | Intron | XP_005260143.1 | ||||
XM_011528326.1 | Intron | XP_011526628.1 |