Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601909 MIM: 615882 | ||||||||||||||||||||
Literature Links: |
GPR21 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GPR21 - G protein-coupled receptor 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005294.2 | 6366 | UTR 5 | NP_005285.1 | |||
XM_005251933.3 | 6366 | UTR 5 | XP_005251990.1 |
RABGAP1 - RAB GTPase activating protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012197.3 | 6366 | Intron | NP_036329.3 | |||
XM_011518440.2 | 6366 | Intron | XP_011516742.1 | |||
XM_011518441.2 | 6366 | Intron | XP_011516743.1 | |||
XM_011518442.2 | 6366 | Intron | XP_011516744.1 | |||
XM_011518444.2 | 6366 | Intron | XP_011516746.1 | |||
XM_017014567.1 | 6366 | Intron | XP_016870056.1 | |||
XM_017014568.1 | 6366 | Intron | XP_016870057.1 | |||
XM_017014569.1 | 6366 | Intron | XP_016870058.1 | |||
XM_017014570.1 | 6366 | Intron | XP_016870059.1 |