Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601909 MIM: 615882 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPR21 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
GPR21 - G protein-coupled receptor 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005294.2 | 287 | Intron | NP_005285.1 | |||
XM_005251933.3 | 287 | UTR 5 | XP_005251990.1 |
RABGAP1 - RAB GTPase activating protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012197.3 | 287 | Intron | NP_036329.3 | |||
XM_011518440.2 | 287 | Intron | XP_011516742.1 | |||
XM_011518441.2 | 287 | Intron | XP_011516743.1 | |||
XM_011518442.2 | 287 | Intron | XP_011516744.1 | |||
XM_011518444.2 | 287 | Intron | XP_011516746.1 | |||
XM_017014567.1 | 287 | Intron | XP_016870056.1 | |||
XM_017014568.1 | 287 | Intron | XP_016870057.1 | |||
XM_017014569.1 | 287 | Intron | XP_016870058.1 | |||
XM_017014570.1 | 287 | Intron | XP_016870059.1 |
Set Membership: |
HapMap Validated |