Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610412 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPTSSB PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SPTSSB - serine palmitoyltransferase small subunit B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040100.1 | 1382 | UTR 3 | NP_001035189.1 | |||
NM_001320679.1 | 1382 | UTR 3 | NP_001307608.1 | |||
XM_017005793.1 | 1382 | UTR 3 | XP_016861282.1 | |||
XM_017005794.1 | 1382 | UTR 3 | XP_016861283.1 |
Set Membership: |
HapMap |