Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTAACAGCTAGGGCCTTGGGGAC[A/G]TGGCAGCAATGCTGCTGTGACCCTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607588 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC107985532 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | |||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LOC107985532 - uncharacterized LOC107985532 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPIL2 - peptidylprolyl isomerase like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317996.1 | Intron | NP_001304925.1 | ||||
NM_014337.3 | Intron | NP_055152.1 | ||||
NM_148175.2 | Intron | NP_680480.1 | ||||
NM_148176.2 | Intron | NP_680481.1 | ||||
XM_005261448.3 | Intron | XP_005261505.1 | ||||
XM_011530041.2 | Intron | XP_011528343.1 | ||||
XM_011530042.2 | Intron | XP_011528344.1 | ||||
XM_011530043.2 | Intron | XP_011528345.1 | ||||
XM_011530044.2 | Intron | XP_011528346.1 | ||||
XM_011530046.2 | Intron | XP_011528348.1 | ||||
XM_011530047.2 | Intron | XP_011528349.1 | ||||
XM_011530049.2 | Intron | XP_011528351.1 | ||||
XM_011530050.2 | Intron | XP_011528352.1 | ||||
XM_011530051.2 | Intron | XP_011528353.1 | ||||
XM_017028705.1 | Intron | XP_016884194.1 | ||||
XM_017028706.1 | Intron | XP_016884195.1 |
Set Membership: |
HapMap Validated |