Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTTGAATTTTAACAACTTCTCAC[A/T]AAGTATCTTTAATCAACACCACTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602963 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC102723704 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC102723704 - uncharacterized LOC102723704 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UBE2D3 - ubiquitin conjugating enzyme E2 D3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300795.1 | Intron | NP_001287724.1 | ||||
NM_003340.6 | Intron | NP_003331.1 | ||||
NM_181886.3 | Intron | NP_871615.1 | ||||
NM_181887.2 | Intron | NP_871616.1 | ||||
NM_181888.3 | Intron | NP_871617.1 | ||||
NM_181889.2 | Intron | NP_871618.1 | ||||
NM_181890.2 | Intron | NP_871619.1 | ||||
NM_181891.2 | Intron | NP_871620.1 | ||||
NM_181892.3 | Intron | NP_871621.1 | ||||
NM_181893.2 | Intron | NP_871622.1 | ||||
XM_017008584.1 | Intron | XP_016864073.1 |