Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614995 MIM: 616012 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL17RE PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IL17RE - interleukin 17 receptor E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193380.1 | Intron | NP_001180309.1 | ||||
NM_153480.1 | Intron | NP_705613.1 | ||||
NM_153481.1 | Intron | NP_705614.1 | ||||
NM_153483.2 | Intron | NP_705616.2 | ||||
XM_006712975.1 | Intron | XP_006713038.1 | ||||
XM_006712976.1 | Intron | XP_006713039.1 | ||||
XM_011533361.2 | Intron | XP_011531663.1 | ||||
XM_011533362.2 | Intron | XP_011531664.1 | ||||
XM_011533363.2 | Intron | XP_011531665.1 | ||||
XM_011533364.2 | Intron | XP_011531666.1 |
JAGN1 - jagunal homolog 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |