Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601982 MIM: 602945 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
OGG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
OGG1 - 8-oxoguanine DNA glycosylase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002542.5 | 1830 | Intron | NP_002533.1 | |||
NM_016819.3 | 1830 | Intron | NP_058212.1 | |||
NM_016820.3 | 1830 | Intron | NP_058213.1 | |||
NM_016821.2 | 1830 | Intron | NP_058214.1 | |||
NM_016826.2 | 1830 | Intron | NP_058434.1 | |||
NM_016827.2 | 1830 | Intron | NP_058436.1 | |||
NM_016828.2 | 1830 | Intron | NP_058437.1 | |||
NM_016829.2 | 1830 | Intron | NP_058438.1 | |||
XM_011533760.1 | 1830 | Intron | XP_011532062.1 | |||
XM_017006493.1 | 1830 | Intron | XP_016861982.1 | |||
XM_017006494.1 | 1830 | Intron | XP_016861983.1 | |||
XM_017006495.1 | 1830 | Intron | XP_016861984.1 | |||
XM_017006496.1 | 1830 | UTR 3 | XP_016861985.1 | |||
XM_017006497.1 | 1830 | Intron | XP_016861986.1 | |||
XM_017006498.1 | 1830 | Intron | XP_016861987.1 | |||
XM_017006499.1 | 1830 | Intron | XP_016861988.1 |
TADA3 - transcriptional adaptor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278270.1 | 1830 | Intron | NP_001265199.1 | |||
NM_006354.3 | 1830 | Intron | NP_006345.1 | |||
NM_133480.2 | 1830 | Intron | NP_597814.1 |
Set Membership: |
HapMap |