Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 128240 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
UTRN PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
UTRN - utrophin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007124.2 | Intron | NP_009055.2 | ||||
XM_005267127.4 | Intron | XP_005267184.1 | ||||
XM_005267130.2 | Intron | XP_005267187.1 | ||||
XM_005267133.2 | Intron | XP_005267190.1 | ||||
XM_006715560.3 | Intron | XP_006715623.1 | ||||
XM_011536101.2 | Intron | XP_011534403.1 | ||||
XM_011536102.2 | Intron | XP_011534404.1 | ||||
XM_011536106.2 | Intron | XP_011534408.1 | ||||
XM_011536107.2 | Intron | XP_011534409.1 | ||||
XM_011536109.2 | Intron | XP_011534411.1 | ||||
XM_017011243.1 | Intron | XP_016866732.1 | ||||
XM_017011244.1 | Intron | XP_016866733.1 | ||||
XM_017011245.1 | Intron | XP_016866734.1 |
Set Membership: |
HapMap |