Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610119 | ||||||||||||||||||||
Literature Links: |
TENM2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TENM2 - teneurin transmembrane protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080428.2 | Intron | NP_001073897.2 | ||||
NM_001122679.1 | Intron | NP_001116151.1 | ||||
XM_005265950.2 | Intron | XP_005266007.1 | ||||
XM_005265952.2 | Intron | XP_005266009.1 | ||||
XM_006714897.2 | Intron | XP_006714960.1 | ||||
XM_011534604.2 | Intron | XP_011532906.1 | ||||
XM_017009660.1 | Intron | XP_016865149.1 | ||||
XM_017009661.1 | Intron | XP_016865150.1 | ||||
XM_017009662.1 | Intron | XP_016865151.1 | ||||
XM_017009663.1 | Intron | XP_016865152.1 | ||||
XM_017009664.1 | Intron | XP_016865153.1 | ||||
XM_017009665.1 | Intron | XP_016865154.1 | ||||
XM_017009666.1 | Intron | XP_016865155.1 | ||||
XM_017009667.1 | Intron | XP_016865156.1 | ||||
XM_017009668.1 | Intron | XP_016865157.1 | ||||
XM_017009669.1 | Intron | XP_016865158.1 |