Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604325 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP2R2B PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PPP2R2B - protein phosphatase 2 regulatory subunit Bbeta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271899.1 | Intron | NP_001258828.1 | ||||
NM_001271900.1 | Intron | NP_001258829.1 | ||||
NM_001271948.1 | Intron | NP_001258877.1 | ||||
NM_181674.2 | Intron | NP_858060.2 | ||||
NM_181675.3 | Intron | NP_858061.2 | ||||
NM_181676.2 | Intron | NP_858062.1 | ||||
NM_181677.2 | Intron | NP_858063.1 | ||||
NM_181678.2 | Intron | NP_858064.1 |
Set Membership: |
HapMap |