Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605584 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DHX38 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DHX38 - DEAH-box helicase 38 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PMFBP1 - polyamine modulated factor 1 binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160213.1 | Intron | NP_001153685.1 | ||||
NM_031293.2 | Intron | NP_112583.2 | ||||
XM_011523357.2 | Intron | XP_011521659.1 | ||||
XM_011523358.2 | Intron | XP_011521660.1 | ||||
XM_011523360.2 | Intron | XP_011521662.1 | ||||
XM_011523361.2 | Intron | XP_011521663.1 | ||||
XM_011523362.2 | Intron | XP_011521664.1 |