Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PHF24 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PHF24 - PHD finger protein 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304333.2 | Intron | NP_001291262.1 | ||||
NM_015297.2 | Intron | NP_056112.1 | ||||
XM_011517827.1 | Intron | XP_011516129.1 | ||||
XM_011517828.2 | Intron | XP_011516130.1 | ||||
XM_011517829.1 | Intron | XP_011516131.1 | ||||
XM_017014553.1 | Intron | XP_016870042.1 | ||||
XM_017014554.1 | Intron | XP_016870043.1 | ||||
XM_017014555.1 | Intron | XP_016870044.1 | ||||
XM_017014556.1 | Intron | XP_016870045.1 |