Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602614 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAP3K7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MAP3K7 - mitogen-activated protein kinase kinase kinase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003188.3 | 3330 | UTR 3 | NP_003179.1 | |||
NM_145331.2 | 3330 | UTR 3 | NP_663304.1 | |||
NM_145332.2 | 3330 | UTR 3 | NP_663305.1 | |||
NM_145333.2 | 3330 | UTR 3 | NP_663306.1 | |||
XM_006715553.2 | 3330 | UTR 3 | XP_006715616.1 | |||
XM_017011226.1 | 3330 | UTR 3 | XP_016866715.1 |
Set Membership: |
HapMap |