Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612752 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CXXC5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CXXC5 - CXXC finger protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317199.1 | Intron | NP_001304128.1 | ||||
NM_001317200.1 | Intron | NP_001304129.1 | ||||
NM_001317201.1 | Intron | NP_001304130.1 | ||||
NM_001317202.1 | Intron | NP_001304131.1 | ||||
NM_001317203.1 | Intron | NP_001304132.1 | ||||
NM_001317204.1 | Intron | NP_001304133.1 | ||||
NM_001317205.1 | Intron | NP_001304134.1 | ||||
NM_001317206.1 | Intron | NP_001304135.1 | ||||
NM_001317207.1 | Intron | NP_001304136.1 | ||||
NM_001317208.1 | Intron | NP_001304137.1 | ||||
NM_001317209.1 | Intron | NP_001304138.1 | ||||
NM_001317210.1 | Intron | NP_001304139.1 | ||||
NM_001317211.1 | Intron | NP_001304140.1 | ||||
NM_016463.8 | Intron | NP_057547.5 | ||||
XM_017009573.1 | Intron | XP_016865062.1 |