Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612752 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CXXC5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CXXC5 - CXXC finger protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317199.1 | Intron | NP_001304128.1 | ||||
NM_001317200.1 | Intron | NP_001304129.1 | ||||
NM_001317201.1 | Intron | NP_001304130.1 | ||||
NM_001317202.1 | Intron | NP_001304131.1 | ||||
NM_001317203.1 | Intron | NP_001304132.1 | ||||
NM_001317204.1 | Intron | NP_001304133.1 | ||||
NM_001317205.1 | Intron | NP_001304134.1 | ||||
NM_001317206.1 | Intron | NP_001304135.1 | ||||
NM_001317207.1 | Intron | NP_001304136.1 | ||||
NM_001317208.1 | Intron | NP_001304137.1 | ||||
NM_001317209.1 | Intron | NP_001304138.1 | ||||
NM_001317210.1 | Intron | NP_001304139.1 | ||||
NM_001317211.1 | Intron | NP_001304140.1 | ||||
NM_016463.8 | Intron | NP_057547.5 | ||||
XM_017009573.1 | Intron | XP_016865062.1 |