Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612752 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CXXC5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CXXC5 - CXXC finger protein 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PSD2 - pleckstrin and Sec7 domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032289.2 | Intron | NP_115665.1 | ||||
XM_011537696.2 | Intron | XP_011535998.1 | ||||
XM_017009975.1 | Intron | XP_016865464.1 | ||||
XM_017009976.1 | Intron | XP_016865465.1 | ||||
XM_017009977.1 | Intron | XP_016865466.1 |