Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602702 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARRDC5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ARRDC5 - arrestin domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080523.1 | 794 | Intron | NP_001073992.1 | |||
XM_005259616.2 | 794 | Intron | XP_005259673.1 | |||
XM_011528181.2 | 794 | Intron | XP_011526483.1 | |||
XM_017027144.1 | 794 | Missense Mutation | AGA,AGC | R,S 177 | XP_016882633.1 | |
XM_017027145.1 | 794 | UTR 3 | XP_016882634.1 |
PLIN3 - perilipin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164189.1 | 794 | UTR 5 | NP_001157661.1 | |||
NM_001164194.1 | 794 | UTR 5 | NP_001157666.1 | |||
NM_005817.4 | 794 | UTR 5 | NP_005808.3 |
Set Membership: |
HapMap |