Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
41 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616035 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FOXJ3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FOXJ3 - forkhead box J3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198850.1 | Intron | NP_001185779.1 | ||||
NM_001198851.1 | Intron | NP_001185780.1 | ||||
NM_001198852.1 | Intron | NP_001185781.1 | ||||
NM_014947.4 | Intron | NP_055762.3 | ||||
XM_005270632.4 | Intron | XP_005270689.1 | ||||
XM_006710458.3 | Intron | XP_006710521.1 | ||||
XM_006710459.3 | Intron | XP_006710522.1 | ||||
XM_011541026.2 | Intron | XP_011539328.1 | ||||
XM_017000693.1 | Intron | XP_016856182.1 | ||||
XM_017000694.1 | Intron | XP_016856183.1 | ||||
XM_017000695.1 | Intron | XP_016856184.1 |