Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602209 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RREB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RREB1 - ras responsive element binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003698.3 | Intron | NP_001003698.1 | ||||
NM_001003699.3 | Intron | NP_001003699.1 | ||||
NM_001003700.1 | Intron | NP_001003700.1 | ||||
NM_001168344.1 | Intron | NP_001161816.1 | ||||
XM_006715157.3 | Intron | XP_006715220.1 | ||||
XM_011514794.2 | Intron | XP_011513096.1 | ||||
XM_011514795.2 | Intron | XP_011513097.1 |