Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
45 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608322 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIF21B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
KIF21B - kinesin family member 21B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252100.1 | 6660 | UTR 3 | NP_001239029.1 | |||
NM_001252102.1 | 6660 | UTR 3 | NP_001239031.1 | |||
NM_001252103.1 | 6660 | UTR 3 | NP_001239032.1 | |||
NM_017596.3 | 6660 | UTR 3 | NP_060066.2 | |||
XM_017000731.1 | 6660 | Intron | XP_016856220.1 | |||
XM_017000732.1 | 6660 | Intron | XP_016856221.1 |
MROH3P - maestro heat like repeat family member 3, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap JSNP Validated |