Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300276 | ||||||||||||||||||||
Literature Links: |
EDA2R PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EDA2R - ectodysplasin A2 receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199687.2 | 845 | Missense Mutation | AAT,ACT | N,T 225 | NP_001186616.1 | |
NM_001242310.1 | 845 | Missense Mutation | AAT,ACT | N,T 246 | NP_001229239.1 | |
NM_001324199.1 | 845 | Missense Mutation | AAT,ACT | N,T 101 | NP_001311128.1 | |
NM_001324201.1 | 845 | Missense Mutation | AAT,ACT | N,T 193 | NP_001311130.1 | |
NM_001324202.1 | 845 | Intron | NP_001311131.1 | |||
NM_001324204.1 | 845 | Intron | NP_001311133.1 | |||
NM_001324205.1 | 845 | Intron | NP_001311134.1 | |||
NM_001324206.1 | 845 | Missense Mutation | AAT,ACT | N,T 227 | NP_001311135.1 | |
NM_021783.4 | 845 | Missense Mutation | AAT,ACT | N,T 225 | NP_068555.1 | |
XM_011531000.1 | 845 | Missense Mutation | AAT,ACT | N,T 246 | XP_011529302.1 | |
XM_011531001.1 | 845 | Missense Mutation | AAT,ACT | N,T 246 | XP_011529303.1 | |
XM_011531002.1 | 845 | Intron | XP_011529304.1 | |||
XM_011531003.2 | 845 | Missense Mutation | AAT,ACT | N,T 166 | XP_011529305.1 | |
XM_011531004.1 | 845 | Intron | XP_011529306.1 | |||
XM_011531005.2 | 845 | Missense Mutation | AAT,ACT | N,T 101 | XP_011529307.1 | |
XM_011531006.2 | 845 | Intron | XP_011529308.1 | |||
XM_011531007.2 | 845 | Intron | XP_011529309.1 | |||
XM_017029703.1 | 845 | Missense Mutation | AAT,ACT | N,T 193 | XP_016885192.1 | |
XM_017029704.1 | 845 | Intron | XP_016885193.1 | |||
XM_017029705.1 | 845 | Missense Mutation | AAT,ACT | N,T 147 | XP_016885194.1 | |
XM_017029706.1 | 845 | Intron | XP_016885195.1 | |||
XM_017029707.1 | 845 | Missense Mutation | AAT,ACT | N,T 122 | XP_016885196.1 | |
XM_017029708.1 | 845 | Intron | XP_016885197.1 |