Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
DENND3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DENND3 - DENN domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014957.2 | 313 | Missense Mutation | AAG,GAG | K,E 17 | NP_055772.2 | |
XM_005250838.4 | 313 | Missense Mutation | AAG,GAG | K,E 30 | XP_005250895.2 | |
XM_005250839.2 | 313 | Missense Mutation | AAG,GAG | K,E 30 | XP_005250896.2 | |
XM_005250840.4 | 313 | Missense Mutation | AAG,GAG | K,E 30 | XP_005250897.2 | |
XM_005250841.3 | 313 | Missense Mutation | AAG,GAG | K,E 30 | XP_005250898.2 | |
XM_005250842.4 | 313 | Missense Mutation | AAG,GAG | K,E 97 | XP_005250899.1 | |
XM_011516933.1 | 313 | Missense Mutation | AAG,GAG | K,E 30 | XP_011515235.1 | |
XM_011516934.2 | 313 | Missense Mutation | AAG,GAG | K,E 30 | XP_011515236.1 | |
XM_011516937.1 | 313 | Missense Mutation | AAG,GAG | K,E 30 | XP_011515239.1 | |
XM_011516938.2 | 313 | Intron | XP_011515240.1 | |||
XM_011516939.2 | 313 | Intron | XP_011515241.1 | |||
XM_011516940.2 | 313 | Intron | XP_011515242.1 | |||
XM_011516941.2 | 313 | Missense Mutation | AAG,GAG | K,E 30 | XP_011515243.1 | |
XM_017013241.1 | 313 | Missense Mutation | AAG,GAG | K,E 30 | XP_016868730.1 | |
XM_017013242.1 | 313 | UTR 5 | XP_016868731.1 | |||
XM_017013243.1 | 313 | Intron | XP_016868732.1 |