Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
81 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612779 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DPYD PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
DPYD - dihydropyrimidine dehydrogenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000110.3 | Intron | NP_000101.2 | ||||
NM_001160301.1 | Intron | NP_001153773.1 | ||||
XM_005270562.3 | Intron | XP_005270619.2 | ||||
XM_006710397.3 | Intron | XP_006710460.1 | ||||
XM_017000507.1 | Intron | XP_016855996.1 | ||||
XM_017000508.1 | Intron | XP_016855997.1 | ||||
XM_017000509.1 | Intron | XP_016855998.1 | ||||
XM_017000510.1 | Intron | XP_016855999.1 |
Set Membership: |
HapMap Validated |