Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603033 MIM: 608315 | ||||||||||||||||||||
Literature Links: |
COLQ PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COLQ - collagen like tail subunit of asymmetric acetylcholinesterase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005677.3 | 1586 | Missense Mutation | TAC,TGC | Y,C 444 | NP_005668.2 | |
NM_080538.2 | 1586 | Missense Mutation | TAC,TGC | Y,C 434 | NP_536799.1 | |
NM_080539.3 | 1586 | Missense Mutation | TAC,TGC | Y,C 410 | NP_536800.2 |
EAF1 - ELL associated factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |