Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604170 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLSCR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PLSCR1 - phospholipid scramblase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021105.2 | Intron | NP_066928.1 | ||||
XM_005247538.3 | Intron | XP_005247595.1 | ||||
XM_011512904.1 | Intron | XP_011511206.1 | ||||
XM_011512905.1 | Intron | XP_011511207.1 | ||||
XM_011512906.2 | Intron | XP_011511208.1 | ||||
XM_011512907.2 | Intron | XP_011511209.1 | ||||
XM_017006628.1 | Intron | XP_016862117.1 | ||||
XM_017006629.1 | Intron | XP_016862118.1 |
Set Membership: |
HapMap |