Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611018 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PAIP2B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PAIP2B - poly(A) binding protein interacting protein 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020459.1 | 313 | Silent Mutation | CAA,CAG | Q,Q 52 | NP_065192.1 | |
XM_005264310.3 | 313 | Silent Mutation | CAA,CAG | Q,Q 52 | XP_005264367.1 | |
XM_005264311.4 | 313 | Silent Mutation | CAA,CAG | Q,Q 52 | XP_005264368.1 | |
XM_011532842.2 | 313 | Silent Mutation | CAA,CAG | Q,Q 73 | XP_011531144.1 | |
XM_011532843.2 | 313 | Silent Mutation | CAA,CAG | Q,Q 52 | XP_011531145.1 |