Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300410 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AMOT PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AMOT - angiomotin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001113490.1 | 4131 | UTR 3 | NP_001106962.1 | |||
NM_133265.2 | 4131 | UTR 3 | NP_573572.1 | |||
XM_005262087.1 | 4131 | UTR 3 | XP_005262144.1 | |||
XM_005262090.1 | 4131 | UTR 3 | XP_005262147.1 | |||
XM_011530875.2 | 4131 | UTR 3 | XP_011529177.1 | |||
XM_017029289.1 | 4131 | UTR 3 | XP_016884778.1 |
MIR4329 - microRNA 4329 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |