Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
41 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZYG11A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZYG11A - zyg-11 family member A, cell cycle regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004339.2 | Intron | NP_001004339.2 | ||||
NM_001307931.1 | Intron | NP_001294860.1 | ||||
XM_011541480.2 | Intron | XP_011539782.1 | ||||
XM_011541481.2 | Intron | XP_011539783.1 | ||||
XM_011541482.2 | Intron | XP_011539784.1 | ||||
XM_011541483.2 | Intron | XP_011539785.1 | ||||
XM_011541484.1 | Intron | XP_011539786.1 | ||||
XM_011541485.1 | Intron | XP_011539787.1 | ||||
XM_011541486.1 | Intron | XP_011539788.1 | ||||
XM_011541487.2 | Intron | XP_011539789.1 | ||||
XM_011541488.1 | Intron | XP_011539790.1 |