Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605198 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PAPD7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PAPD7 - PAP associated domain containing 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171805.1 | Intron | NP_001165276.1 | ||||
NM_001171806.1 | Intron | NP_001165277.1 | ||||
NM_006999.4 | Intron | NP_008930.1 | ||||
XM_005248234.3 | Intron | XP_005248291.1 | ||||
XM_011513945.1 | Intron | XP_011512247.1 | ||||
XM_017008988.1 | Intron | XP_016864477.1 | ||||
XM_017008989.1 | Intron | XP_016864478.1 | ||||
XM_017008990.1 | Intron | XP_016864479.1 |
Set Membership: |
Validated |