Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGCCCTGTAGCAGAAGTACTTAG[C/T]GCTTTGCATTCTGCGCGCCACCCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602872 MIM: 603382 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLIC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CLIC1 - chloride intracellular channel 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MSH5 - mutS homolog 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002441.4 | Intron | NP_002432.1 | ||||
NM_025259.5 | Intron | NP_079535.4 | ||||
NM_172165.3 | Intron | NP_751897.1 | ||||
NM_172166.3 | Intron | NP_751898.1 |
MSH5-SAPCD1 - MSH5-SAPCD1 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |