Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616990 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLUL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLUL1 - clusterin like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289036.1 | Intron | NP_001275965.1 | ||||
NM_001318522.1 | Intron | NP_001305451.1 | ||||
NM_014410.4 | Intron | NP_055225.1 | ||||
NM_199167.1 | Intron | NP_954636.1 | ||||
XM_005258103.3 | Intron | XP_005258160.1 | ||||
XM_006722322.2 | Intron | XP_006722385.1 | ||||
XM_011525648.2 | Intron | XP_011523950.1 | ||||
XM_011525649.1 | Intron | XP_011523951.1 | ||||
XM_011525651.2 | Intron | XP_011523953.1 | ||||
XM_011525652.1 | Intron | XP_011523954.1 | ||||
XM_011525653.2 | Intron | XP_011523955.1 | ||||
XM_017025707.1 | Intron | XP_016881196.1 |