Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610593 MIM: 607858 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAP6D1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAP6D1 - MAP6 domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PARL - presenilin associated rhomboid like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037639.2 | Intron | NP_001032728.1 | ||||
NM_001324436.1 | Intron | NP_001311365.1 | ||||
NM_001324437.1 | Intron | NP_001311366.1 | ||||
NM_001324438.1 | Intron | NP_001311367.1 | ||||
NM_018622.6 | Intron | NP_061092.3 | ||||
XM_005247582.4 | Intron | XP_005247639.1 | ||||
XM_017006800.1 | Intron | XP_016862289.1 | ||||
XM_017006801.1 | Intron | XP_016862290.1 | ||||
XM_017006802.1 | Intron | XP_016862291.1 | ||||
XM_017006803.1 | Intron | XP_016862292.1 |