Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCGCCTTCCGAGGCCAGGTGAAC[G/T]TTACCGGCCCCCTGACCTCCAGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604069 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100126584 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LOC100126584 - uncharacterized LOC100126584 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TPD52L1 - tumor protein D52-like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003395.2 | 311 | UTR 5 | NP_001003395.1 | |||
NM_001003396.2 | 311 | Intron | NP_001003396.1 | |||
NM_001003397.2 | 311 | Intron | NP_001003397.1 | |||
NM_001292026.2 | 311 | UTR 5 | NP_001278955.1 | |||
NM_001300994.2 | 311 | Intron | NP_001287923.1 | |||
NM_001318903.1 | 311 | Intron | NP_001305832.1 | |||
NM_001318907.1 | 311 | Intron | NP_001305836.1 | |||
NM_003287.3 | 311 | Intron | NP_003278.1 | |||
XM_005267122.2 | 311 | Intron | XP_005267179.1 | |||
XM_006715557.3 | 311 | Intron | XP_006715620.1 | |||
XM_017011237.1 | 311 | Intron | XP_016866726.1 | |||
XM_017011238.1 | 311 | Intron | XP_016866727.1 | |||
XM_017011239.1 | 311 | UTR 5 | XP_016866728.1 | |||
XM_017011240.1 | 311 | Intron | XP_016866729.1 |