Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114090 MIM: 606832 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAST PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ERAP1 - endoplasmic reticulum aminopeptidase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040458.1 | 5169 | Intron | NP_001035548.1 | |||
NM_001198541.1 | 5169 | Intron | NP_001185470.1 | |||
NM_016442.3 | 5169 | UTR 3 | NP_057526.3 | |||
XM_005272015.4 | 5169 | Intron | XP_005272072.1 | |||
XM_005272016.4 | 5169 | Intron | XP_005272073.1 | |||
XM_011543480.2 | 5169 | Intron | XP_011541782.1 | |||
XM_011543481.2 | 5169 | Intron | XP_011541783.1 | |||
XM_011543484.2 | 5169 | Intron | XP_011541786.1 | |||
XM_011543485.2 | 5169 | Intron | XP_011541787.1 | |||
XM_011543486.2 | 5169 | Intron | XP_011541788.1 | |||
XM_017009580.1 | 5169 | Intron | XP_016865069.1 | |||
XM_017009581.1 | 5169 | Intron | XP_016865070.1 | |||
XM_017009582.1 | 5169 | UTR 3 | XP_016865071.1 | |||
XM_017009583.1 | 5169 | Intron | XP_016865072.1 |
Set Membership: |
HapMap |