Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609268 | ||||||||||||||||||||
Literature Links: |
SREK1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SREK1 - splicing regulatory glutamic acid and lysine rich protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077199.2 | Intron | NP_001070667.1 | ||||
NM_001270492.1 | Intron | NP_001257421.1 | ||||
NM_001323527.1 | Intron | NP_001310456.1 | ||||
NM_001323529.1 | Intron | NP_001310458.1 | ||||
NM_001323533.1 | Intron | NP_001310462.1 | ||||
NM_001323534.1 | Intron | NP_001310463.1 | ||||
NM_001323535.1 | Intron | NP_001310464.1 | ||||
NM_139168.3 | Intron | NP_631907.1 | ||||
XM_011543171.2 | Intron | XP_011541473.1 | ||||
XM_017009055.1 | Intron | XP_016864544.1 | ||||
XM_017009056.1 | Intron | XP_016864545.1 |