Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607703 | ||||||||||||||||||||
Literature Links: |
NUP210 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NUP210 - nucleoporin 210 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024923.3 | 5614 | Silent Mutation | GCA,GCC | A,A 1883 | NP_079199.2 | |
XM_005264994.3 | 5614 | Intron | XP_005265051.1 | |||
XM_011533538.2 | 5614 | Intron | XP_011531840.1 | |||
XM_017006018.1 | 5614 | Silent Mutation | GCA,GCC | A,A 1011 | XP_016861507.1 | |
XM_017006019.1 | 5614 | Silent Mutation | GCA,GCC | A,A 1000 | XP_016861508.1 | |
XM_017006020.1 | 5614 | Silent Mutation | GCA,GCC | A,A 988 | XP_016861509.1 | |
XM_017006021.1 | 5614 | Intron | XP_016861510.1 |