Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 182350 MIM: 600283 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP1A3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ATP1A3 - ATPase Na+/K+ transporting subunit alpha 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GRIK5 - glutamate ionotropic receptor kainate type subunit 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301030.1 | 4621 | UTR 3 | NP_001287959.1 | |||
NM_002088.4 | 4621 | Intron | NP_002079.3 | |||
XM_005258821.3 | 4621 | Intron | XP_005258878.1 | |||
XM_011526862.2 | 4621 | Intron | XP_011525164.1 | |||
XM_011526863.2 | 4621 | UTR 3 | XP_011525165.1 | |||
XM_011526865.1 | 4621 | UTR 3 | XP_011525167.1 | |||
XM_011526867.1 | 4621 | UTR 3 | XP_011525169.1 | |||
XM_011526868.1 | 4621 | UTR 3 | XP_011525170.1 | |||
XM_011526869.2 | 4621 | Intron | XP_011525171.1 | |||
XM_011526870.2 | 4621 | Intron | XP_011525172.1 | |||
XM_011526871.2 | 4621 | Intron | XP_011525173.1 | |||
XM_017026713.1 | 4621 | UTR 3 | XP_016882202.1 |
Set Membership: |
HapMap |