Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611223 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKT3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
AKT3 - AKT serine/threonine kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206729.1 | Intron | NP_001193658.1 | ||||
NM_005465.4 | Intron | NP_005456.1 | ||||
NM_181690.2 | Intron | NP_859029.1 | ||||
XM_005272994.4 | Intron | XP_005273051.1 | ||||
XM_005272995.2 | Intron | XP_005273052.1 | ||||
XM_006711726.3 | Intron | XP_006711789.1 | ||||
XM_011544012.2 | Intron | XP_011542314.1 | ||||
XM_011544013.2 | Intron | XP_011542315.1 | ||||
XM_011544014.2 | Intron | XP_011542316.1 | ||||
XM_016999985.1 | Intron | XP_016855474.1 |
Set Membership: |
Validated |