Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611966 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRAPPC9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TRAPPC9 - trafficking protein particle complex 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160372.3 | 4639 | UTR 3 | NP_001153844.1 | |||
NM_001321646.1 | 4639 | UTR 3 | NP_001308575.1 | |||
NM_031466.7 | 4639 | UTR 3 | NP_113654.4 | |||
XM_011517326.2 | 4639 | Intron | XP_011515628.1 | |||
XM_011517328.2 | 4639 | Intron | XP_011515630.1 | |||
XM_011517330.2 | 4639 | UTR 3 | XP_011515632.1 | |||
XM_017013893.1 | 4639 | Intron | XP_016869382.1 | |||
XM_017013894.1 | 4639 | UTR 3 | XP_016869383.1 |